Learn More
Bullard™ RT Series Tychem™ Respirator Hoods
SDP
Catalog No. 17989620 Shop All Bullard Company Products
Change view
Click to view available options
Material:
Tychem QC
Tychem SL
Includes:
Inflatable neck cuff
Sport neck cuff
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Includes
17-989-620 Tychem SL Inflatable neck cuff
19-168-315 Tychem QC Inflatable neck cuff
19-321-718 Tychem QC Sport neck cuff
19-321-719 Tychem SL Sport neck cuff
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 17-989-620 Supplier Bullard™ Supplier No. RT2
Add to Cart
Add to Cart
This item is not returnable. View return policy
RT hoods are available in two different Tychem materials and are held in place by the inflatable neck cuff (RT1 and RT2 models) or a sport material neck cuff (RT3 and RT4 models).
  • Tychem™ QC (quality coated) is a lightweight, supple material coated with polyethylene to provide splash protection against many acids, bases, and other liquid chemicals
  • Tychem SL (Saranex™ laminate) is coated with Saranex 23-P to provide protection from an even wider range of chemicals
  • RT1/RT3 = Tychem QC
  • RT2/RT4 = Tychem SL
  • Panoramic view minimizes claustrophobic feeling sometimes experienced in other respirator hoods
  • Sport or inflatable neck cuff design provides positive custom seal
  • Three-channel air delivery system promotes curvature of hood for better fit
  • Long inner and outer bibs for splash protection
  • Extra-large acetate lens
  • No suspension is required
  • Breathing tube connects using a sleeve on the back of the hood and a clamp
Order Info
Lens cover available. (See Cat. No. 17-986-288)
Specifications
Assigned Protection Factor (APF) 1,000
Material Tychem SL
Product Type Respirator Hood
For Use With (Application) Industrial environments, petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing
For Use With (Equipment) PA30, EVA, and/or RT Airline respiraor
Includes Inflatable neck cuff
Model RT2
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.