Learn More
Agilent Technologies™ RTCA Software Basic Single License
Description
- xCELLigence RTCA Software Pro analyzes real-time data, generates diverse plot types and automatically calculates dose response curves and parameters such as % cytolysis, IC50 or KT50
- Quickly evaluate efficacy of different treatments with automated normalization and compensation features
- Calculate slope, Cell Index doubling time, % of control, and EC50/ IC50
- Support for FDA 21 CFR Part 11 compliance to ensure the authenticity and integrity of electronic data
Specifications
Specifications
| Type | Impedance Software |
| For Use With (Equipment) | xCELLigence RTCA Analyzer |