Learn More
Agilent Technologies RTCA Software Virology Single License
Catalog No. 10010025
Change view
Click to view available options
Type:
Software
Catalog No. Type
10-010-025 Software
1 options
Catalog No. 10-010-025 Supplier Agilent Technologies Supplier No. S280790089
Add to Cart
Add to Cart
The RTCA Software Virology Single License for Real-Time Cell Analysis (RTCA) Software Pro streamlines traditional viral titer methods, including TCID50 and VRNT, by monitoring viral cytopathic effects (CPE) over time.
  • The Virology Module is an add-on module for either RTCA Software Pro Basic or RTCA Software Pro IMT.
  • It streamlines traditional viral titer methods, including TCID50 and VRNT, by monitoring viral cytopathic effects (CPE) over time with customizable calculation methods and user-defined CPE thresholds.
  • Monitor viral Cytopathic Effects (CPE) in real time without the use of labels
  • Obtain deeper Insight into Viral Dynamics as mediated by CPE to support advanced virology research and process development
  • Automate calculation of 50% tissue culture infectious dose (TCID50) and Virus Reduction Neutralization test (VRNT)
  • Includes customizable calculation methods and user-defined CPE thresholds
  • Measure viral titer and virus neutralizing activity in an easy-to-use assay with higher sensitivity and shorter hands-on time
  • Built-in quality control and higher objectivity of interpretation compared to labor-intensive imaging methods
Specifications
Type Software
For Use With (Equipment) xCELLigence RTCA Pro Software
Includes Single License