Learn More
Omega Engineering, Inc. RTD PROBE W/INSULATE WIRE TUBE
Supplier:  Omega Engineering, Inc. PR10E31001/812
Encompass
AccuracyClass A from -100°C to 450°C (-148 to 842°F); Class B otherwise, per IEC 60751Cold-End Max TemperatureLow (L);High (H) and Extreme (E), 250°C (482°F)DimensionsStandard diameters of 1/8”, 3/16”, and 1/4” ; variable lengthsIngress ProtectionIP67 (with exception of “High” (10H) type)Nominal Resistance CurveChoice of PT100Ω or PT1000ΩProcess Temperature Range-200 to 600°C (-328 to 1112°F)Protection headNoneWire Length / Terminations40” standard (longer lengths available)Omega's RTD probe with insulated lead wire has a shrink-tube strain relief support at the sheath/wire transition to provide wire flexibility and durability. This 316 stainless steel probe is offered in 2-, 3-, or 4-wire configuration. This is available in three temperature ranges covering -200°C to 600°C (-328 to 1112°F) to accommodate your specific temperature needs. Applications for this probe include liquid immersion, slurries and emulsions, granul
Catalog No. NC2742919
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.