Learn More
Omega Engineering, Inc. RTD Probe with Insulated Wire and Shrink-Tube Support
Supplier:  Omega Engineering, Inc. PR10L31001/86
Encompass
RTD Probe with Insulated Wire and Shrink-Tube SupportLow temperature range: -50 to 250°C (-58 to 482°F)High temperature range: -50 to 450°C (-58 to 842°F)Extreme temperature range: -200 to 600°C (-328 to 1112°F)Class A accuracy conforming to IEC 60751
Catalog No. NC2734305