Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RTN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583716
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583716 Supplier Invitrogen™ Supplier No. PA583716
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SITPPSSGTE PSAAESQGKG SISEDELITA IKEAKGLSYE TAENPRPVGQ LADRPEVKAR SGPPTIPSPL DHEASSAESG DSEIELVS Highest antigen sequence indentity to the following orthologs: Rat - 85%, Mouse - 86%.

Reticulon-1 was formerly designated NSP for Neuroendocrine-Specific-Protein, because it is specifically expressed in neural and neuroendocrine tissues. The NSP-gene has been mapped by fluorescence in situ hybridization to human chromosome 14q21-q22. The NSP-gene encodes three overlapping proteins, i.e. reticulon-1A (NSP-A), reticulon-1B (NSP-B), and reticulon-1C (NSP-C). These proteins were found to be anchored to membranes of the endoplasmic reticulum through their common carboxy-terminal regions. Reticulon-1A is a protein with a molecular weight of approximately 135 kDa, which occurs in various isoforms presumably depending on the degree of phosphorylation of serine residues. In lung cancer diagnosis Reticulon-1A appeared to be a reliable marker for the detection of neuroendocrine differentiation, since most of the small cell lung carcinoma (SCLC) and carcinoid tumors showed expression of Reticulon-1A. Reticulon-1B is a phosphoprotein with a MW of 45 kDa and is restricted to the lung cancer cell line NCI-H82. Reticulon-1B is so far not found in human tissues. Reticulon-1C is a protein with a MW of 23 kDa which is not phosphorylated and is found with Reticulon-1A in SCLC (cell lines) and not in non-SCLC (cell cultures).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RTN1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RTN1
Gene Accession No. Q16799
Gene Alias 0710005K15Rik; 4930441F12Rik; I79_024631; LOC100750648; MGC133250; MGC40985; Neuroendocrine-specific protein; Nsp; R75279; reticulon 1; reticulon-1; RP23-450G14.1; RTN1; Rtn1-a; Rtn1-b; Rtn1-c; S-rex
Gene Symbols RTN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RTN1. Recombinant protein control fragment (Product #RP-102963).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6252
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.