Learn More
Invitrogen™ RTN4RL2 Recombinant Rabbit Monoclonal Antibody (P14)

Description
Specificity: The antibody binds to human NgR2 with a Kd of 2.3 nM and to rat NgR2 with a Kd of 0.48 nM (bivalent binding).
Specifications
Specifications
| Antigen | RTN4RL2 |
| Applications | ELISA, Flow Cytometry, Immunoprecipitation, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | P14 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene | RTN4RL2 |
| Gene Accession No. | Q7M6Z0, Q80WD1, Q86UN3 |
| Gene Alias | ngR2; NGRH1; NGRL3; Nogo receptor-like 3; Nogo-66 receptor homolog 1; Nogo-66 receptor-related protein 2; reticulon 4 receptor-like 2; reticulon-4 receptor-like 2; RGD:727797}; Rtn4rl2; rtn4rl2 {ECO:0000312 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.