Learn More
Med Vet International Miltex Jansen Retractor 4 25in 2-3/8in Spread 3 X 3 Blunt Prongs 10mm Deep German 11-2
Supplier:  Med Vet International MIL112
Encompass_Preferred
Miltex Jansen Retractor 4 25in 2-3/8in Spread 3 X 3 Blunt Prongs 10mm Deep German 11-2
Catalog No. 50-307-0268
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.