Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ru(OAc)2[(R)-tolbinap], TCI America™
SDP

Catalog No. R01681G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R01681G 1 g
R0168200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R01681G Supplier TCI America Supplier No. R01681G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 116128-29-1
Molecular Formula C52H48O4P2Ru
Molecular Weight (g/mol) 899.97
MDL Number MFCD09753021,MFCD09753021
InChI Key OXESSJMVRGFBNX-UHFFFAOYSA-N
Synonym Diacetato[(R)-(+)-2,2′C-bis(di-p-tolylphosphino)-1,1′C-binaphthyl]ruthenium(II)
PubChem CID 85364800
IUPAC Name bis(acetic acid) {2'-[bis(4-methylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(4-methylphenyl)phosphane ruthenium
SMILES [Ru].CC(O)=O.CC(O)=O.CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1

Specifications

Color Black-Yellow
Quantity 1 g
Formula Weight 897.95
Physical Form Crystalline Powder
Chemical Name or Material Ru(OAc)2[(R)-tolbinap]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.