Learn More
DWK Life Sciences Rubber Conical Gasket 8-Piece Set Guko, from EPDM, conical rubber gaskets, for filtering flasks
SDP
Catalog No. 09841086 Shop All DWK Life Sciences Products
Change view
Click to view available options
8 pieces, size 22 to 84
Catalog No.
09-841-086 8 pieces, size 22 to 84
1 options
Catalog No. 09-841-086 Supplier DWK Life Sciences Supplier No. 292020001
Add to Cart
Add to Cart
The rubber sleeve is required to achieve gas tightness during vacuum filtration.
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Buechner funnels are cylinders with an integrated large-pored sieve plate. Without additional filter paper, only coarse filtration is possible. Buechner funnels can also be used in vacuum filtration. The applied vacuum accelerates the filtration.
  • For tight closure in vacuum filtration applications
  • Conical rubber gaskets
  • for filtering flasks
TRUSTED_SUSTAINABILITY
Specifications
Description 8 Gukos, size 22 to 84
Material EPDM
For Use With (Application) Sealing; Filtering
No. per Pack 1/Pk.
Autoclavable Autoclavable