Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Rubber O-Ring
SCI_ED

Catalog No. S88416 Shop All Eisco Products
Change view
Click to view available options
Size:
14/23
19/26
24/29
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
S88416 24/29
S88414 14/23
S88415 19/26
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S88416 Supplier Eisco™ Supplier No. CH1038C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eisco™ Rubber O-Ring is available in a variety of standard joint sizes.

  • Replacement rubber O-rings
  • Available in a variety of standard joint sizes
  • Helps provide a tight leak proof seal with screw caps

Specifications

For Use With (Application) Seal caps
For Use With (Equipment) Glassware and labware
Quantity 1 Ea.
Product Type O-Ring
Material Rubber
Color Black
Diameter (Metric) Outside 25.7 mm
Diameter (English) Outside 1.01 in.
Size 24/29
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.