Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Rubber Pipet Filler
SCI_ED

Catalog No. S23644 Shop All Eisco Products
Change view
Click to view available options
Number of Channels:
3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Number of Channels
S23644 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S23644 Supplier Eisco™ Supplier No. CH0580A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rubber Pipet Filler is a bulb-type filler for pipetting noxious solutions and microbial suspensions.

  • Comprises bulb and three pinch valves
  • Helps control influx and efflux of pipette contents

Specifications

Autoclavable Not Autoclavable
Color Red
Disposable Single-use
For Use With Pipet Filler
Grip Type Single-handed
Material Rubber
Model Standard
Number of Channels 3
Sterility Non-sterile
Type Pipette Filler
Volume (Metric) 60 mL
For Use With (Application) For pipetting noxious solutions and microbial suspensions
Includes Bulb and 3 pinch valves
Diameter (Metric) Outer 54 mm dia. bulb
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.