Description
Convenient scrapers with angle-shaped edge
- Scrape precipitate from container
- Hard rubber with angled edges
- Use section of glass rod as handle
Specifications
Specifications
| Product Type | Policemen Scraper |
| For Use With (Equipment) | 0.48 cm dia. Glass Rod |
| For Use With (Application) | Designed to scrape precipitate from container by using section of Glass rod as handle |
| Includes | Angle shaped edge |
| Material | Rubber |
| Handle Type | Glass Rod (sold separately) |