Learn More
Rubbermaid™ Microfiber Finish System
Catalog No. 18367029 Shop All Newell Rubbermaid Inc Products
Change view
Click to view available options
Dimensions (L x W):
18 x 5 in.
Catalog No. Dimensions (L x W)
18-367-029 18 x 5 in.
1 options
This item is not returnable. View return policy
Catalog No. 18-367-029 Supplier Rubbermaid™ Supplier No. FGQ050000000
Add to Cart
Add to Cart
This item is not returnable. View return policy
Enables controlled absorption and release of finish
  • Delivers 200 to 300 square feet of coverage with single mop load of finish
  • Split microfiber for fast and smooth finish application
  • Smooth, nonporous bucket for easy cleaning
  • Includes bucket with lid, sieve, 8 pads, 1 frame
Specifications
Product Type Finish System
Dimensions (L x W) 18 x 5 in.
Format Split microfiber
Packaging Quantity 1/Pk.