Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam RuBi-5-HT, Caged-serotonin Compound, Solid, 10 mg
Description
- Molecular weight: 736.64 g/mol
- Form: Solid
- Properties: Highly water-soluble caged-serotonin (5-HT) compound based on ruthenium photochemistry
- Activation: Excitable with visible wavelengths and releases serotonin after one- or two-photon excitation
- Performance: High quantum efficiency allows for use at low concentrations; enables photoactivation of 5-HT receptors with visible or two-photon light sources for single-cell or single-spine precision
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Serotonin |
| Shipping Condition | Ambient |
| Kit Contents | 10 mg RuBi-5-HT |
| Buffer | 100% RuBi-5-HT |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Formulation | 100% RuBi-5-HT |
| Quantity | 10 mg |
| For Use With (Application) | Photoactivation of 5-HT receptors |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?