Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RuBi-Dopa, Tocris Bioscience™

Catalog No. 493210
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-3210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 49-3210 Supplier R&D Systems Supplier No. 4932/10
This item is not returnable. View return policy

Caged dopamine; exhibits two-photon sensitivity

RuBi-caged dopamine (Cat. No. 3548); exhibits two-photon sensitivity. Enables non-invasive, optical activation of dopamine receptors with the spatial resolution of a single dendritic spine. Does not display phototoxicity at a concentration of 300 μM.

Chemical Identifiers

Molecular Formula C31H36Cl2N5O2PRu
Synonym (bis(2,2'-Bipyridine-N,N')trimethylphosphine)-(S)-(3,4-Dihydroxyphenyl)ethylamino ruthenium(2+) dichloride complex

Specifications

Quantity 10 mg
Formula Weight 713.6
Percent Purity >98%
Chemical Name or Material RuBi-Dopa
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.