Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubidium Chloride, Metal min. 99%., MP Biomedicals

Catalog No. ICN10282550 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10282550 50 g
ICN10282505 5 g
ICN10282510 10 g
ICN10282580 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10282550 Supplier MP Biomedicals, Inc Supplier No. 0210282550
Add to Cart
Add to Cart

Solutions may be slightly hazy

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11-9-7791
Molecular Formula ClRb
Molecular Weight (g/mol) 120.92
MDL Number MFCD00011187
InChI Key FGDZQCVHDSGLHJ-UHFFFAOYSA-M
Synonym rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
PubChem CID 62683
ChEBI CHEBI:78672
IUPAC Name rubidium(1+) chloride
SMILES [Cl-].[Rb+]

Specifications

Quantity 50 g
Assay Percent Range Metal min. 99%.
Formula Weight 120.9
Chemical Name or Material Rubidium Chloride
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.