Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubidium molybdenum oxide, 99.5% (Metals basis), Thermo Scientific™

Catalog No. AA3951609
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3951609 10 g
AA3951604 2 g
AA3951618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3951609 Supplier Thermo Scientific Chemicals Supplier No. 3951609

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13718-22-4
Molecular Formula Rb2MoO4
MDL Number MFCD00049518
Synonym Rubidium molybdate

Specifications

Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Soluble in water.
Formula Weight 330.88
Odor Odorless
Appearance White powder
Chemical Name or Material Rubidium molybdenum oxide
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 237-271-1
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.