Learn More
AdipoGen Rubpy2-5-ClAPA-PF62
SDP
Supplier:  AdipoGen CDXR0028M010
Encompass_Preferred
Chemical. CAS 204273-42-7. Formula C34H26F12ClN7OP2Ru. MW 975.0679. Synthetic. Reactive probe containing s fluorescent metal-ligand complex for the determination of nucleotide base sequences.
Catalog No. 50-256-0126
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.