Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[RuCl(p-cymene)((S)-segphos(regR))]Cl, TCI America™
SDP

Catalog No. R01551G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R01551G 1 g
R0155200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R01551G Supplier TCI America Supplier No. R01551G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 944451-29-0
Molecular Formula C48H42Cl2O4P2Ru
Molecular Weight (g/mol) 916.778
MDL Number MFCD09753017
InChI Key HZLSGDCLBWZKRB-UHFFFAOYSA-L
Synonym Chloro[(S)-(-)-5,5′C-bis(diphenylphosphino)-4,4′C-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride
PubChem CID 72376325
IUPAC Name dichlororuthenium;[4-(5-diphenylphosphanyl-1,3-benzodioxol-4-yl)-1,3-benzodioxol-5-yl]-diphenylphosphane;1-methyl-4-propan-2-ylbenzene
SMILES CC1=CC=C(C=C1)C(C)C.C1OC2=C(O1)C(=C(C=C2)P(C3=CC=CC=C3)C4=CC=CC=C4)C5=C(C=CC6=C5OCO6)P(C7=CC=CC=C7)C8=CC=CC=C8.Cl[Ru]Cl

Specifications

Quantity 1 g
Formula Weight 916.78
Physical Form Crystalline Powder
Chemical Name or Material [RuCl(p-cymene)((S)-segphos(regR))]Cl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.