Learn More
Apexbio Technology LLC Rufinamide 106308-44-5 25mg
SDP
Supplier:  Apexbio Technology LLC B228025
Encompass_Preferred
Rufinamide is a small-molecule inhibitor targeting voltage-gated sodium channels It is designed to modulate neuronal excitability by stabilizing sodium channel inactivation thereby reducing repetitive neuronal firing and seizure propagation Rufinamide exerts its biological activity primarily through inhibition of voltage-gated sodium channels Based on these pharmacological properties Rufinamide holds research potential in neuroscience studies particularly as an adjunctive therapeutic candidate for severe epilepsy subtypes such as Lennox-Gastaut syndrome and in investigations of cellular electrophysiological mechanisms underlying epileptic disorders
Catalog No. 50-000-90500
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.