Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RUNDC3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569851
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569851 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569851 Supplier Invitrogen™ Supplier No. PA569851
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 92%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

RUNDC3B is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUNDC3B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Rundc3b
Gene Accession No. Q96NL0
Gene Alias Gm440; Rap2 binding protein 9; Rap2-binding protein 9; Rap2-interacting protein 9; RPIB9; RPIP9; RPIP-9; RUN domain containing 3B; RUN domain-containing protein 3B; rundc3b; zgc:101561
Gene Symbols Rundc3b
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human RUNDC3B.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154661
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.