Learn More
Invitrogen™ RUNX1/RUNX1T1 Polyclonal Antibody

Description
RUNX1/RUNX1T1 Polyclonal Antibody for Western Blot, ChIP, ChIP-seq, ELISA
Specifications
Specifications
| Antigen | RUNX1/RUNX1T1 |
| Applications | ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | whole serum with 0.05% sodium azide |
| Gene | RUNX1 |
| Gene Accession No. | Q01196, Q06455 |
| Gene Alias | acute myelogenous leukemia 1 translocation 1, cyclin-D related; acute myeloid leukemia 1 protein; AML1; aml1 oncogene; AML1-EVI-1; AML1-EVI-1 fusion protein; AML1-MTG8; AML1T1; AMLCR1; CBF2alpha; CBFA2; CBFA2T1; CBFA2T1 isoform r1t1-11a61; CBFA2T1 isoform r1t1-11a62; CBFA2T1 isoform r1t1-11a63; CBFA2T1 isoform r1t1-11a64; CBFA2T1 isoform r1t1-11a65; CBFA2T1 isoform r1t1-7a47; CBFA2T1 isoform r1t1-7a48; CBFA2T1 isoform r1t1-7a49; CBFA2T1 isoform r1t1-7a50; CBFA2T1 isoform r1t1-7a51; CBFA2T1 isoform r1t1-7a52; CBFA2T1 isoform r1t1-7d53; CBFA2T1 isoform r1t1-7d54; CBFA2T1 isoform r1t1-7d55; CBFA2T1 isoform r1t1-7d56; CBFA2T1 isoform r1t1-8a57; CBFA2T1 isoform r1t1-8a58; CBFA2T1 isoform r1t1-8a59; CBFA2T1 isoform r1t1-8a60; CBF-alpha-2; CDR; Core-binding factor subunit alpha-2; core-binding factor, runt domain, alpha subunit 2; core-binding factor, runt domain, alpha subunit 2; translocated to, 1; cyclin D-related; Cyclin-D-related protein; Eight twenty one protein; ETO; EVI-1; MTG8; myeloid translocation gene on 8q22; oncogene AML-1; PEA2-alpha B; PEBP2aB; PEBP2alpha; PEBP2-alpha B; polyomavirus enhancer-binding protein 2 alpha B subunit; Protein CBFA2T1; Protein ETO; Protein MTG8; runt related transcription factor 1; runt related transcription factor 1; translocated to, 1 (cyclin D related); runt-related transcription factor 1; runt-related transcription factor 1; translocated to, 1 (cyclin D-related); RUNX1; RUNX1 translocation partner 1; RUNX1/ZNF687 fusion; RUNX1T1; SL3/AKV core-binding factor alpha B subunit; SL3-3 enhancer factor 1 alpha B subunit; transcription factor; zinc finger MYND domain-containing protein 2; ZMYND2 |
| Gene Symbols | RUNX1, RUNX1T1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.