Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RUNX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582787 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582787 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582787 Supplier Invitrogen™ Supplier No. PA582787
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LNSAPSPFNP QGQSQITDPR QAQSSPPWSY DQSYPSYLSQ MTSPSIHSTT PLSSTRGTGL PAITDVPRRI SGASELGPFS DPRQFPSISS LTESRFSNPR MHYPA Highest antigen sequence indentity to the following orthologs: Rat - 81%, Mouse - 100%.

RUNX2, a nuclear protein, is a member of the RUNX family of transcription factors and has an Runt DNA-binding domain. It is essential for membranous and endochondral bone formation. It not only regulates osteoblastic differentiation and skeletal morphogenesis but also acts as a scaffold for nucleic acids and regulatory factors involved in skeletal gene expression. The protein can bind DNA both as a monomer and with more affinity, as a subunit of a heterodimeric complex. It plays a critical role in increasing TGFBR1 expression by osteoblasts and functions in cooperation with DLX5 or a related factor to activate osteoblast-specific gene expression. Mutations lead to disorders in bone development like cleidocranial dysplasia (CCD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUNX2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RUNX2
Gene Accession No. Q13950
Gene Alias Acute myeloid leukemia 3 protein; AKV core binding factor; Aml3; AML-3; Cbf; Cbfa1; Cbfa-1; CBF-alpha-1; CCD; CCD1; CLCD; core binding factor alpha 1; core binding factor alpha 1 subunit; core-binding factor subunit alpha-1; core-binding factor, runt domain, alpha subunit 1; LOW QUALITY PROTEIN: runt-related transcription factor 2; LS3; Oncogene AML-3; Osf2; OSF-2; osteoblast-specific transcription factor 2; PEA2aA; PEA2-alpha A; PEBP2 alpha A; PEBP2A; Pebp2a1; PEBP2aA; PEBP2-alpha A; Pebpa2a; Polyomavirus enhancer-binding protein 2 alpha A subunit; runt domain, alpha subunit 1; runt related transcription factor 2; runt-related transcription factor 2; RUNX 2; RUNX family transcription factor 2; Runx2; RUNX-2; SL3/AKV core-binding factor alpha A subunit; SL3-3 enhancer factor 1 alpha A subunit; transcription factor RUNX2; transcription factor RUNX2/CBFA1
Gene Symbols RUNX2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RUNX2. Recombinant protein control fragment (Product #RP-102296).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 860
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.