Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RUNX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120837
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120837 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120837 Supplier Invitrogen™ Supplier No. PA5120837
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: MCF7, HeLa, Mouse testis. The target is usually found in the following locations: Nucleus.

RUNX2, a nuclear protein, is a member of the RUNX family of transcription factors and has an Runt DNA-binding domain. It is essential for membranous and endochondral bone formation. It not only regulates osteoblastic differentiation and skeletal morphogenesis but also acts as a scaffold for nucleic acids and regulatory factors involved in skeletal gene expression. The protein can bind DNA both as a monomer and with more affinity, as a subunit of a heterodimeric complex. It plays a critical role in increasing TGFBR1 expression by osteoblasts and functions in cooperation with DLX5 or a related factor to activate osteoblast-specific gene expression. Mutations lead to disorders in bone development like cleidocranial dysplasia (CCD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUNX2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene RUNX2
Gene Accession No. Q08775, Q13950, Q9Z2J9
Gene Alias Acute myeloid leukemia 3 protein; AKV core binding factor; Aml3; AML-3; Cbf; Cbfa1; Cbfa-1; CBF-alpha-1; CCD; CCD1; CLCD; core binding factor alpha 1; core binding factor alpha 1 subunit; core-binding factor subunit alpha-1; core-binding factor, runt domain, alpha subunit 1; LOW QUALITY PROTEIN: runt-related transcription factor 2; LS3; Oncogene AML-3; Osf2; OSF-2; osteoblast-specific transcription factor 2; PEA2aA; PEA2-alpha A; PEBP2 alpha A; PEBP2A; Pebp2a1; PEBP2aA; PEBP2-alpha A; Pebpa2a; Polyomavirus enhancer-binding protein 2 alpha A subunit; runt domain, alpha subunit 1; runt related transcription factor 2; runt-related transcription factor 2; RUNX 2; RUNX family transcription factor 2; Runx2; RUNX-2; SL3/AKV core-binding factor alpha A subunit; SL3-3 enhancer factor 1 alpha A subunit; transcription factor RUNX2; transcription factor RUNX2/CBFA1
Gene Symbols RUNX2
Host Species Rabbit
Immunogen Synthetic peptide
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12393, 367218, 860
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.