Learn More
Apexbio Technology LLC Tie2 kinase inhibitor 948557-43-5 100mg
SDP
Supplier:  Apexbio Technology LLC A5979100
Encompass_Preferred
Tie2 kinase inhibitor (CAS 948557-43-5) is a selective and reversible small-molecule inhibitor targeting the Tie2 receptor tyrosine kinase a critical modulator of endothelial cell survival angiogenesis and vascular integrity By inhibiting Tie2 autophosphorylation this compound disrupts subsequent signaling pathways involved in endothelial maturation and tumor-associated angiogenesis In vitro studies demonstrate potent inhibition of Tie2 kinase activity (IC50 0 25 M) with substantial selectivity over other kinases including p38 In vivo treatment reduces angiogenesis and tumor progression in murine xenograft tumor models This inhibitor is thus relevant for research examining angiogenic mechanisms and cancer progression
Catalog No. 50-000-62450
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.