Learn More
TARGETMOL CHEMICALS INC SHMT-IN-2 5MG
Supplier: TARGETMOL CHEMICALS INC T624045MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SHMT-IN-2 (compound 2) is a specific human SHMT1/2 small molecule inhibitor that acts on SHMT1 (IC50 13 nM) and SHMT2 (IC50 66 nM). SHMT-IN-2 inhibits the growth of many human cancer cells and is sensitive to B-cell lymphoma. purity: 98%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.