Learn More
Selleck Chemical LLC Timolol Maleate S4123-50mg
SDP
Supplier:  Selleck Chemical LLC S412350MG
Encompass_Preferred
Timolol Maleate (MK-950 (S)-Timolol Maleate) is a non-selective beta-adrenergic receptor antagonist for 1/ 2 with Ki of 1 97 nM/2 0 nM
Catalog No. 50-310-9353
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.