Learn More
Invitrogen™ Ruplizumab Recombinant Human Monoclonal Antibody (hu5c8 (Ruplizumab))

Description
Specificity: The antibody binds to CD40L with an IC50 of 0.845 μg/mL.
Specifications
Specifications
| Antigen | Ruplizumab |
| Applications | Flow Cytometry, Inhibition Assays, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | hu5c8 (Ruplizumab) |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene | Ntrk1 |
| Gene Accession No. | P04629 |
| Gene Alias | BG-9588; hu5c8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.