Learn More
ALADDIN SCIENTIFIC CORPORATION Mitomycin C, 25mg
Supplier:  ALADDIN SCIENTIFIC CORPORATION M47663325MG
Encompass_Preferred
Mitomycin C is a systemic chemotherapy compound and an antibiotic obtained fromStreptomyces caespitosus It is the first bioreductive alkylating agent to be identified and is a derivative of 3-amino-5-hydroxybenzoic acid (AHBA) D-glucosamine L-methionine and carbamoyl phosphate Chemical structure- AziridineMitomycin C fromStreptomyces caespitosushas been used for the treatment of feeder layers such as PMEF (primary mouse embryonic fibroblasts) and CD1 mouse embryonic fibroblasts (MEFs) for the culture of hESCs (human embryonic stem cells) It has also been used for the treatment of BLC (basal-like cancer) cell line max 216 359 nm
Catalog No. 50-000-82534
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.