Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ruthenium(IV) oxide hydrate, Ru 54% min

Catalog No. AA1180303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1180303 1 g
AA1180306 5 g
AA1180314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1180303 Supplier Thermo Scientific Chemicals Supplier No. 01180303
Add to Cart

CAS: 32740-79-7 | O2Ru | 133.07 g/mol

OxidationRuthenium(IV) oxide hydrate is used as a starting material for the preparation of ruthenium tetraoxide by oxidation. It is also used for the coating of titanium anodes and for the preparation of resistors. It is an active material in supercapacitor due to its high charge transfer capability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32740-79-7
Molecular Formula O2Ru
Molecular Weight (g/mol) 133.07
MDL Number MFCD00149846
InChI Key WOCIAKWEIIZHES-UHFFFAOYSA-N
IUPAC Name dioxoruthenium
SMILES O=[Ru]=O

Specifications

Melting Point 1200° (sublimation)
Color Black
Physical Form Powder
Quantity 1 g
Assay Percent Range Ru 54% min.
Solubility Information Insoluble in water. Soluble in HClInsoluble in water.
Formula Weight 133.07 (Anhydrous)
Odor Odorless
Appearance Black powder
Chemical Name or Material Ruthenium(IV) oxide hydrate
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P501c
DOTInformation DOT Class: 5.1, Packaging Group: II
EINECSNumber 234-840-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.