Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKA™ RV 10 Digital with Dry Ice Condenser
SDP

Catalog No. 11116993 Shop All IKA Works Products
Change view
Click to view available options
Capacity (Metric):
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
11-116-993 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-116-993 Supplier IKA™ Supplier No. 0010005210
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Represents the continual enhancement of the IKA RV 10 rotary evaporator line

  • 4 L heating bath
  • Universal water/oil heating bath
  • Coated dry ice condenser ideal for the distillation of low-boiling solvents
  • Motorized lift with 'safety-stop' function
  • RS 232 and IR Interface
  • Optimized heating bath volume for fast heat-up
  • Highly efficient 1500 cm2 condenser unit
  • Rugged design
  • Ergonomically placed handles on Heating bath
  • Smooth start
  • Left-right interval
  • Timer function
  • Dry-run protection
  • Safety temperature circuits
  • Lower-end stop
  • Push-off mechanism to loosen tight fitted glassware
  • Easy and safe operation due to ergonomical design of the user interface

Specifications

Capacity (Metric) 1 L
Speed 5 to 280 rpm
Temperature Range (Metric) Ambient to 180°C
Type Rotary Evaporator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.