Learn More
MARSHALL SCIENTIFIC Edwards RV12 Rotary Vane Vacuum Pump
Supplier: MARSHALL SCIENTIFIC EDA65501903
The RV series of small oil sealed rotary vane pumps deliver excellent ultimate vacuum pressure, high pumping speeds and superior vapour handling capabilities with quiet operation. With over 300,000 units produced, these pumps offer proven performance that sets the industry standard for R&D and scientific pumping applications. All RV pumps are approved to UL and CSA standards by an external test house and feature our unique mode selector switch, meaning one model is suitable for both high vacuum or high throughput applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.