Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RVG29-Cys

Catalog No. 89035
Change view
Click to view available options
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
89-035 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035 Supplier Tocris Bioscience™ Supplier No. 8903/5
Add to Cart
Add to Cart

Nicotinic acetylcholine receptor targeting peptide for neuronal delivery

RVG29-Cys is a peptide derived from rabies virus glycoprotein (RVG) able to specifically bind on the nicotinic acetylcholine receptors (nAchR) present on neuronal cells. RVG29-Cys can be used to functionalize LNPs via thiol-maleimide click chemistry for targeting nAchR present on blood-brain barrier (BBB). mRNA LNPs functionalized with RVG29-Cys shows improved transfection efficiency in cultured brain endothelial and neuronal cells, and in vivo, after systemic administration in mice.

Specifications

Purity 95%
CAS 1404289-25-3
Content And Storage Store at -20°C
Molecular Weight (g/mol) Observed: 3369.79
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.