Learn More
Cayman Chemical RW3trIfluoroacetate salt 10mg
SDP
Supplier:  Cayman Chemical 3749310 MG
Encompass_Preferred
An antimicrobial peptide; active against E. coli and S. aureus (IC50s = 16 and 8 µM, respectively); induces hemolysis of isolated ovine erythrocytes
Catalog No. 50-291-6085