Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RWDD3 Monoclonal Antibody (OTI8B3), TrueMAB™, OriGene
SDP

Catalog No. 501684229
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4229 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4229 Supplier OriGene Supplier No. TA811754
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RWDD3 (RSUME), a small RWD-containing protein, has a central role in sumoylation by enhancing SUMO conjugation in the regulatory network of immune-inflammatory signals. RWDD3 increases Ikappa-Beta sumoylation and stability. In addition, RWDD3 inhibits TNF-a-induced kappaB-LUC (Luciferase) reporter activity, showing the functional consequence of Ikappa-Beta increased stability. RSUME-enhanced sumoylation of Ikappa-Beta leads to the inhibition of NF-kappa-Beta activity on two well-known inflammatory genes, IL-8 and cyclooxygenase-2 (Cox-2) and therefore may also favor anti-inflammatory pathways. Expression of RWDD3 was induced under hypoxic conditions and it has a potential role during vascularization. Both BMP-4 and RWDD3 may be interesting targets for inhibiting steps involved in pituitary tumorigenesis.

Specifications

Antigen RWDD3
Applications Western Blot
Classification Monoclonal
Clone OTI8B3
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene RWDD3
Gene Accession No. Q9Y3V2
Gene Alias RSUME
Gene Symbols RWDD3
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-267 of human RWDD3 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25950
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.