Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RWDD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112258
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112258 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112258 Supplier Invitrogen™ Supplier No. PA5112258
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specificity: Human RWDD4.

TRUSTED_SUSTAINABILITY

Specifications

Antigen RWDD4
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene RWDD4
Gene Accession No. Q6NW29
Gene Alias BC016198; FAM28A; family with sequence similarity 28, member A; Gm1942; Protein FAM28A; RWD domain containing 4; RWD domain containing 4A; RWD domain-containing protein 4; RWD domain-containing protein 4A; Rwdd4; Rwdd4a
Gene Symbols RWDD4
Host Species Rabbit
Immunogen E. coli-derived Human RWDD4A/RWDD4 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201965
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.