Learn More
Invitrogen™ RXFP4 Polyclonal Antibody

Description
The antibody detects endogenous level of total RXFP4 / GPR100 protein.
Specifications
Specifications
| Antigen | RXFP4 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 2.30 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | RXFP4 |
| Gene Accession No. | Q7TQP4, Q8TDU9 |
| Gene Alias | G protein-coupled receptor 100; GPCR142; GPR100; G-protein coupled receptor 100; G-protein coupled receptor GPCR142; Insulin-like peptide INSL5 receptor; JEG62; MGC126556; MGC126558; relaxin 3 receptor 2; relaxin family peptide receptor 4; relaxin/insulin like family peptide receptor 4; relaxin/insulin-like family peptide receptor 4; Relaxin-3 receptor 2; relaxin-3/INSL7 receptor 2; RLN3 receptor 2; Rln3r2; Rxfp4; RXFPR4; SALPR |
| Gene Symbols | RXFP4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.