Learn More
Med Vet International Pancreplus Tablets, 500ct.
Supplier: Med Vet International RXPANCREPLUST5
A pancreatic enzyme concentrate of porcine origin fortified with vitamins A, D and E. It is used as a digestive aid in replacement therapy where digestion of protein, carbohydrate and fat is inadequate due to exocrine pancreatic insufficiency. For use in dogs and cats. Each tablet contains a minimum: Lipase, 9,000 USP units Protease, 57,000 USP units Amylase, 64,000 USP units Vitamin A, 1,000 IU Vitamin D3, 100 IU Vitamin E, 10 IU
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.