Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RY655 Mouse Anti-Rat CD18 (Integrin β2), BD OptiBuild™

Catalog No. BDB773645 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB773645 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB773645 Supplier BD Biosciences Supplier No. 773645
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

CD18 (Integrin β2) Monoclonal Antibody Specifically detects CD18 (Integrin β2) in Rat Samples. It is validated for Flow Cytometry.

The WT.3 monoclonal antibody specifically recognizes the 95-100 kDa β2 subunit (CD18), which forms heterodimers with CD11 α integrin subunits (CD11a or αL, CD11b or αM, CD11c or αX) to create LFA-1, Mac-1, and gp150, 95. This antibody has demonstrated function-blocking activity in in vitro assays measuring LFA-1 binding to ICAM-1 (CD54) and has been reported to inhibit leukocyte infiltration in an in vivo system. The BD Horizon RealYellow 655 (RY655) Dye is a yellow-green tandem fluorochrome with an excitation maximum at 557 nm and an emission maximum at 654 nm. RY655 can be used on both spectral and conventional cytometers and is designed for excitation by the Yellow-Green laser (561-nm).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD18 (Integrin β2)
Applications Flow Cytometry
Classification Monoclonal
Clone WT.3
Concentration 0.2 mg/mL
Conjugate RY655
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Integrin β2 chain; integrin beta-2; Itgb2
Host Species Mouse
Immunogen PHA-stimulated rat splenocytes and rat thymic lymphoma FTL-43
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 309684
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.