Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RY655 Rat Anti-Mouse CD153 (TNFSF8), BD OptiBuild™

Catalog No. BDB772451 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB772451 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB772451 Supplier BD Biosciences Supplier No. 772451
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

CD153 (TNFSF8) Monoclonal Antibody Specifically detects CD153 (TNFSF8) in Mouse Samples. It is validated for Flow Cytometry.

The RM153 monoclonal antibody recognizes a 40-kDa type-II membrane glycoprotein, CD30 ligand (CD30L or CD153), part of the NGF/TNF superfamily. This antibody stimulates B-cell proliferation, antigen-specific antibody production, and polyclonal immunoglobulin secretion in the presence of cytokines, and costimulates activated T cell proliferation. CD153 is expressed on activated T cells (predominantly CD4+), activated EL4 cells, and activated Th1 and Th2 cells. The RM153 monoclonal antibody inhibits CD153 binding to CD30. It is conjugated with BD Horizon RealYellow 655 (RY655) Dye, a yellow-green tandem fluorochrome with an excitation maximum at 557 nm and an emission maximum at 654 nm. RY655 can be used on both spectral and conventional cytometers, designed for excitation by the 561-nm Yellow-Green laser. It serves as an alternative to PE-Cy5 on conventional instruments with a Yellow-Green laser, with a recommended optical filter centered near 660 nm (e.g., 660/30-nm or 670/30-nm bandpass filter). On conventional and spectral instruments with both Blue (488-nm) and Yellow-Green (561-nm) lasers and appropriate detectors, it can be used with RB670.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD153 (TNFSF8)
Applications Flow Cytometry
Classification Monoclonal
Clone RM153
Concentration 0.2 mg/mL
Conjugate RY655
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CD30 Ligand; Cd30l; CD30LG; Tnfsf8
Host Species Rat
Immunogen CHO cells transfected with mouse CD153 gene (Tnfsf8)
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21949
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.