Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RY743 Mouse Anti-Human EphJ2, BD OptiBuild™

Catalog No. BDB773581 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB773581 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB773581 Supplier BD Biosciences Supplier No. 773581
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

EphJ2 Monoclonal Antibody Specifically detects EphJ2 in Human Samples. It is validated for Flow Cytometry.

The 371805 monoclonal antibody specifically recognizes Ephrin type-A receptor 2 (EphJ2). EphJ2 is a ~130 kDa type I transmembrane glycoprotein encoded by EPHJ2, belonging to the ephrin receptor subfamily within the receptor tyrosine kinase (RTK) family. It's also known as Epithelial cell kinase (ECK) or Epithelial cell receptor protein tyrosine kinase. Its extracellular region includes an N-terminal globular domain, a cysteine-rich domain, and 2 fibronectin type III repeats, while the cytoplasmic domain has two tyrosine residues that act as major autophosphorylation sites and a kinase domain. EphJ2 is expressed by epithelial cells, dendritic cells, Langerhans cells, keratinocytes, and endothelial cells. It functions as a receptor for glycophosphatidylinositol (GPI) membrane-linked Ephrin-A family members (A1-A5) and is involved in regulating cellular growth, adhesion, migration, survival, and angiogenesis. Its expression can be upregulated on vascular endothelium in certain breast, prostate, and colon cancers, and on some metastatic tumor cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphJ2
Applications Flow Cytometry
Classification Monoclonal
Clone 371805
Concentration 0.2 mg/mL
Conjugate RY743
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias EPHJ2; ECK; ARCC2; CTPA; CTPP1; CTRCT6
Host Species Mouse
Immunogen Human EphJ2 Recombinant Protein
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1969
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.