Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-1,3-Butanediol, 98%

Catalog No. AC217970050
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC217970050 5 g Glass bottle
AC217970010 1 g Glass bottle
AC217970250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC217970050 Supplier Thermo Scientific Chemicals Supplier No. 217970050
Add to Cart

CAS: 24621-61-2 | C4H10O2 | 90.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24621-61-2
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00064278
InChI Key PUPZLCDOIYMWBV-BYPYZUCNSA-N
Synonym s-+-1,3-butanediol, s-butane-1,3-diol, 3s-butane-1,3-diol, s-1,3-butanediol, s-+-butane-1,3-diol, 1,3-butanediol, 3s, bu2, 1,3-butanediol, s, pubchem6722
PubChem CID 446973
ChEBI CHEBI:52688
IUPAC Name (3S)-butane-1,3-diol
SMILES CC(CCO)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 109.0°C (14.0 mmHg)
Flash Point 121°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4385 to 1.4405
Linear Formula CH3CH(OH)CH2CH2OH
Quantity 5 g
Specific Gravity 1
Specific Rotation + 30.00
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone, methyl ethyl ketone, ethanol, slightly soluble in ether, practically insoluble in aliphatic hydrocarbons,, benzene, toluene and ccl4
Formula Weight 90.12
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material (S)-(+)-1, 3-Butanediol
Show More Show Less
EINECSNumber 246-363-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.