Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-1-(4-Bromophenyl)ethylamine 98.0+%, TCI America™
SDP

Catalog No. B367425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B367425G 25 g
B36745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B367425G Supplier TCI America Supplier No. B367425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27298-97-1
Molecular Formula C8H11BrN
Molecular Weight (g/mol) 201.09
MDL Number MFCD00066026
InChI Key SOZMSEPDYJGBEK-LURJTMIESA-O
Synonym s---1-4-bromophenyl ethylamine, s-1-4-bromophenyl ethanamine, s---4-bromo-alpha-phenylethylamine, s-1-4-bromophenyl ethylamine, 1s-1-4-bromophenyl ethanamine, s-1-4-bromophenyl-ethylamine, 1s-1-4-bromophenyl ethan-1-amine, s---1-4-bromo-phenyl-ethylamine, benzenemethanamine,4-bromo-a-methyl-, as, s-1-4-bromophenyl ethanamine-hcl
PubChem CID 852999
IUPAC Name (1S)-1-(4-bromophenyl)ethan-1-aminium
SMILES C[C@H]([NH3+])C1=CC=C(Br)C=C1

Specifications

Color Yellow
Quantity 25 g
UN Number 2735
Formula Weight 200.08
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (S)-(-)-1-(4-Bromophenyl)ethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.