Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-1-BOC-3-aminopyrrolidine, 95%

Catalog No. AC435450010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC435450010 1 g Glass bottle
AC435450050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC435450010 Supplier Thermo Scientific Chemicals Supplier No. 435450010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 147081-44-5 | C9H18N2O2 | 186.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147081-44-5
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.26
MDL Number MFCD03419271
InChI Key CMIBWIAICVBURI-UHFFFAOYNA-N
Synonym s-1-boc-3-aminopyrrolidine, s-3-amino-1-n-boc-pyrrolidine, s---1-boc-3-aminopyrrolidine, tert-butyl 3s-3-aminopyrrolidine-1-carboxylate, s-tert-butyl 3-aminopyrrolidine-1-carboxylate, s-3-amino-n-boc-pyrrolidine, s-bocap, s---1-tert-butoxycarbonyl-3-aminopyrrolidine, s-3-amino-1-boc-pyrrolidine, s---n-boc-3-aminopyrrolidine
PubChem CID 854071
IUPAC Name tert-butyl (3S)-3-aminopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(N)C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 120.0°C to 125.0°C (5.0 Torr)
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation -1.5° to -3.0° (20°C, 589nm) (C=1, chloroform)
Formula Weight 186.25
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material (S)-(-)-1-BOC-3-aminopyrrolidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes serious eye damage.
Risk of explosion if heated under confinement.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.