Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(S)-1-Cyclopropylethylamine, ChiPros™, 98%, ee 98+%
Click to view available options
Quantity:
1 g
5 g
Description
(S)-1-Cyclopropylethylamine acts as a chiral and organic building block in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-1-Cyclopropylethylamine acts as a chiral and organic building block in organic synthesis.
Solubility
Miscible with water.
Notes
Air sensitive. Incompatible with acids and strong oxidizing agents.
(S)-1-Cyclopropylethylamine acts as a chiral and organic building block in organic synthesis.
Solubility
Miscible with water.
Notes
Air sensitive. Incompatible with acids and strong oxidizing agents.
Specifications
Specifications
| Melting Point | -68°C |
| Density | 0.799 |
| Boiling Point | 95°C |
| Flash Point | 3°C (37°F) |
| Quantity | 1 g |
| UN Number | UN2733 |
| Sensitivity | Air Sensitive |
| Synonym | s-1-cyclopropylethylamine, s-1-cyclopropylethanamine, 1s-1-cyclopropylethanamine, 1s-1-cyclopropylethan-1-amine, 1 s-1-cyclopropylethanamine, n-s-1-cyclopropyl-ethyl-amine, s-1-cyclopropylethylamine, chipros, cyclopropanemethanamine,a-methyl-, as, s-1-cyclopropylethylamine, chipros, ee 98+%, s-1-cyclopropylethylamine, chipros r , produced by basf |
| Solubility Information | Miscible with water. |
| Formula Weight | 85.15 |
| Show More |
Safety and Handling
| Hazard Category | H225-H314-H335 |
| Hazard Statement | GHS H Statement H225-H314-H318 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction