Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-1-Methoxy-2-propylamine, ChiPros 98+%, ee 96+%, Thermo Scientific™

Catalog No. AAL1632503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1632503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1632503 Supplier Thermo Scientific Chemicals Supplier No. L1632503

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(S)-(+)-1-Methoxy-2-propylamine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Fully miscible in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. It is sensitive to air. Store under dry inert gas. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 99636-32-5
Melting Point -95°C
Density 0.844
Boiling Point 98°C to 99°C
Flash Point 9°C (48°F)
Molecular Formula C4H11NO
Refractive Index 1.4044
Quantity 1 g
MDL Number MFCD01075730
UN Number UN2733
Beilstein 5725728
Sensitivity Air Sensitive
Synonym s-+-1-methoxy-2-propylamine, 2-propanamine, 1-methoxy-, 2s, 2s-1-methoxypropan-2-amine, s-2-amino-1-methoxypropane, s-1-methoxypropan-2-amine, s-1-methoxy-2-propylamine, s-1methoxy-2-aminopropane, s-1-methoxy-2-propanamine, s-1-methoxy-2-aminopropane, s-2-amino-3-methoxypropane
Solubility Information Fully miscible in water.
InChI Key NXMXETCTWNXSFG-BYPYZUCNSA-N
Optical Rotation +11.5° (Neat)
SMILES CC(COC)N
IUPAC Name (2S)-1-methoxypropan-2-amine
Molecular Weight (g/mol) 89.138
PubChem CID 7021536
Formula Weight 89.14
Percent Purity ≥98%
Assay ee ≥96%
Chemical Name or Material (S)-(+)-1-Methoxy-2-propylamine
Show More Show Less
Hazard Category H225-H302-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318-H302
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 700-220-3
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.