Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-1-Phenyl-1,3-propanediol, 98%

Catalog No. AC417760050
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC417760050 5 g Glass bottle
AC417762500 250 mg Glass bottle
AC417760010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC417760050 Supplier Thermo Scientific Chemicals Supplier No. 417760050
Add to Cart

CAS: 96854-34-1 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96854-34-1
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00221510
InChI Key RRVFYOSEKOTFOG-VIFPVBQESA-N
Synonym 1s-1-phenylpropane-1,3-diol, s-1-phenyl-1,3-propanediol, s-1-phenylpropane-1,3-diol, 1s-phenyl-1,3-propanediol, s-1-phenyl-propane-1,3-diol, s, ?-1-phenyl-1,3-propanediol, 1,3-propanediol,1-phenyl-, 1s, 1,3-propanediol, 1-phenyl-, 1s, s---1-phenyl-1,3-propanediol, puriss sum of enantiomers, gc
PubChem CID 6950537
IUPAC Name (1S)-1-phenylpropane-1,3-diol
SMILES C1=CC=C(C=C1)C(CCO)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 62.0°C to 66.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation Condition −54.50° (22°C c=1,1,2-dichloroethane)
Specific Rotation − 54.50
Formula Weight 152.19
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material (S)-1-Phenyl-1, 3-propanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.