Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-1-Phenylethyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. P079625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P079625G 25 g
P07961G 1 g
P07965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P079625G Supplier TCI America Supplier No. P079625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1445-91-6
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00064264
InChI Key WAPNOHKVXSQRPX-ZETCQYMHSA-N
Synonym s-1-phenylethanol, s---1-phenylethanol, 1s-1-phenylethanol, 1s-1-phenylethan-1-ol, unii-2mic4qly2x, 1-phenylethanol, s, 2mic4qly2x, s-1-phenethyl alcohol, s-1-phenyl-1-ethanol, s-1-phenylethan-1-ol
PubChem CID 443135
ChEBI CHEBI:16346
IUPAC Name (1S)-1-phenylethan-1-ol
SMILES C[C@H](O)C1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 98°C
Quantity 25 g
UN Number 2937
Formula Weight 122.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(-)-1-Phenylethyl Alcohol
EINECSNumber (3)-1012&(3)-1049
RTECSNumber DO9275000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.