Learn More
R&D Systems™ S-100 Fraction Conjugation Kit, HeLa Cells

Description
Specifications
Specifications
| Description | S-100 Proteasome Fraction |
| For Use With (Application) | Supports controlled conjugation of exogenously added or endogenously contained substrate proteins through the ubiquitin proteasome system; |
| Quantity | 25 Tests |
| Storage Requirements | Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.